missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyraldehyde dimethyl acetal, 98+%

Product Code. 11474517
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474517 5 g 5g
11484517 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474517 Supplier Thermo Scientific Alfa Aesar Supplier No. B21072.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19060-15-2 | C6H16NO2 | 134.20 g/mol

4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.

Solubility
Miscible with benzene.

Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19060-15-2
Molecular Formula C6H16NO2
Molecular Weight (g/mol) 134.20
MDL Number MFCD00189373
InChI Key TYVAXMOICMBSMT-UHFFFAOYSA-O
Synonym 4-aminobutyraldehyde dimethyl acetal, 4,4-dimethoxybutylamine, 4,4-dimethoxy-1-butanamine, 4-aminobutyraldehyde dimethylacetal, acmc-1bqj6, 4-aminobutyraldehyde dimethyl, 1-amino-4,4-dimethoxybutane, cs-waa0041, tyvaxmoicmbsmt-uhfffaoysa, 4-aminobutyraldehydedimethylacetal
PubChem CID 5463803
SMILES COC(CCC[NH3+])OC

Specifications

Boiling Point 85°C (22 mmHg)
Flash Point 59°C (138°F)
Linear Formula H2N(CH2)3CH(OCH3)2
Refractive Index 1.43
Quantity 5 g
UN Number UN2734
Sensitivity Air Sensitive
Solubility Information Miscible with benzene.
IUPAC Name 4,4-dimethoxybutan-1-aminium
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material 4-Aminobutyraldehyde dimethyl acetal
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Aminobutyraldehyde dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-Aminobutyraldehydedimethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.