missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 97%

Product Code. 11478843
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478843 2500 g 2500g
11458843 100 g 100g
11468843 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478843 Supplier Thermo Scientific Alfa Aesar Supplier No. A11016.0E
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 56-12-2 | C4H9NO2 | 103.121 g/mol

In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point ∼195°C (decomposition)
Linear Formula H2N(CH2)3CO2H
Beilstein 906818
Merck Index 14,430
Solubility Information Soluble in water.
Quantity 2500 g
Odor Odorless
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material 4-Aminobutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.