missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric acid, MedChemExpress

Product Code. 16458726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458726 Supplier Medchem Express Supplier No. HYB1306
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 4-Aminohippuric acid is a diagnostic agent, useful in medical tests involving the kidney, used in the measurement of renal plasma flow.

4-Aminohippuric acid is a diagnostic agent, useful in medical tests involving the kidney, used in the measurement of renal plasma flow.

Specifications

Chemical Name or Material 4-Aminohippuric acid
CAS 61-78-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H10N2O3
Quantity 100 mg
Synonym p-Aminohippuric acid
Solubility Information DMSO : 50 mg/mL (257.48 mM; Need ultrasonic) ∣H2O : ≥ 1.7 mg/mL (8.75 mM)
SMILES O=C(O)CNC(C1=CC=C(N)C=C1)=O
Molecular Weight (g/mol) 194.19
Formula Weight 194.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.