missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyl-1-Boc-4-hydroxypiperidine, 97%

Product Code. 15459568
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459568 5 g 5g
15469568 250 mg 250mg
15449568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459568 Supplier Thermo Scientific Alfa Aesar Supplier No. H63079.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 392331-66-7 | C11H22N2O3 | 230.308 g/mol

4-Aminomethyl-1-Boc-4-hydroxypiperidine can be used in agrochemical, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminomethyl-1-Boc-4-hydroxypiperidine can be used in agrochemical, pharmaceutical and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392331-66-7
Molecular Formula C11H22N2O3
Molecular Weight (g/mol) 230.308
MDL Number MFCD11579803
InChI Key XYWCDAFPRBDRER-UHFFFAOYSA-N
PubChem CID 23396319
IUPAC Name tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)(CN)O

Specifications

Color White
Quantity 5 g
Sensitivity Air Sensitive
Synonym tert-butyl 4-aminomethyl-4-hydroxypiperidine-1-carboxylate, 1-boc-4-aminomethyl-4-hydroxypiperidine, tert-butyl 4-aminomethyl-4-hydroxytetrahydro-1 2h-pyridinecarboxylate, 4-aminomethyl-1-boc-4-hydroxypiperidine, 4-aminomethyl-1-boc-piperidin-4-ol, tert-butyl 4-aminomethyl-4-hydroxypiperidinecarboxylate, tert-butyl 4-aminomethyl-4-hydroxy-piperidine-1-carboxylate, 4-aminomethyl-4-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl4-aminomethyl-4-hydroxypiperidine-1-carboxylate
Solubility Information Slightly soluble in water.
Formula Weight 230.3
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Aminomethyl-1-Boc-4-hydroxypiperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.