missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzoic Acid, TRC

Product Code. 30135323
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135323 10 g 10g
30136665 100 g 100g
17903805 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135323 Supplier Toronto Research Chemicals Supplier No. TRCA61523010G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-91-7
Molecular Formula C8 H9 N O2
Molecular Weight (g/mol) 151.16
Synonym 4-Aminomethylbenzoic Acid, Tranexamic Acid Imp. D (EP), Benzoic acid, 4-(aminomethyl)-, p-Toluic acid, α-amino- (6CI,8CI), 4-(Aminomethyl)benzoic acid, 4-Carboxybenzylamine, Benzylamine-4-carboxylic acid, Gumbix, NSC 41629, PAMBA, Styptopur, p-(Aminomethyl)benzoic acid, p-Carboxybenzylamine, α-Amino-p-toluic acid
IUPAC Name 4-(aminomethyl)benzoic acid
SMILES NCc1ccc(cc1)C(=O)O

Specifications

InChI Formula InChI=1S/C8H9NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5,9H2,(H,10,11)
Formula Weight 151.0633
Percent Purity >95%
Quantity 10 g
Chemical Name or Material 4-Aminomethylbenzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.