missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)pyrimidine hydrochloride, 97%

Product Code. 15485327
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485327 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485327 Supplier Thermo Scientific Alfa Aesar Supplier No. H32582.ME
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1138011-17-2 | C5H7N3 | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1138011-17-2
Melting Point 176°C to 178°C
Molecular Formula C5H7N3
MDL Number MFCD06796202,MFCD07375466
Quantity 500 mg
Synonym 4-aminomethyl pyrimidine, 4-pyrimidinemethanamine, 1-pyrimidin-4-yl methanamine, 4-aminomethylpyrimidine, pyrimidin-4-ylmethyl amine dihydrochloride, pyrimidin-4-yl methanamine, c-pyrimidin-4-yl-methylamine, 4-pyrimidinemethanamine 9ci, 1-pyrimidin-4-ylmethanamine dihydrochloride
InChI Key AUHXBTKGPUVFCB-UHFFFAOYSA-N
SMILES NCC1=NC=NC=C1
IUPAC Name pyrimidin-4-ylmethanamine
Molecular Weight (g/mol) 109.13
PubChem CID 12933348
Formula Weight 145.59
Percent Purity 97%
Chemical Name or Material 4-(Aminomethyl)pyrimidine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.