missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethylbenzenesulfonamide Hydrochloride, TRC

Product Code. 30346608
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346608 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346608 Supplier Toronto Research Chemicals Supplier No. TRCA6307985G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Mafenide Hydrochloride
CAS 138-37-4
Molecular Formula C7 H10 N2 O2 S . Cl H
Quantity 5 g
Synonym Benzenesulfonamide, 4-(aminomethyl)-, hydrochloride (1:1), Benzenesulfonamide, 4-(aminomethyl)-, monohydrochloride (9CI), p-Toluenesulfonamide, α-amino-, hydrochloride (6CI), p-Toluenesulfonamide, α-amino-, monohydrochloride (8CI), 4-(Aminomethyl)benzene sulfonamide monohydrochloride, 4-Aminosulfonylbenzylamine hydrochloride, 4-Homosulfanilamide hydrochloride, Homosulfamine, Homosulfamine hydrochloride, Homosulfanilamide hydrochloride, Mafenide hydrochloride, Marfanil, Sulfamylon hydrochloride, p-(Aminomethyl)benzenesulfonamide hydrochloride, p-(Aminomethyl)benzenesulfonamide monohydrochloride, p-Aminomethylphenylsulfonamide hydrochloride, α-Amino-p-toluenesulfonamide hydrochloride
InChI Formula InChI=1S/C7H10N2O2S.ClH/c8-5-6-1-3-7(4-2-6)12(9,10)11;/h1-4H,5,8H2,(H2,9,10,11);1H
SMILES Cl.NCc1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-(aminomethyl)benzenesulfonamide;hydrochloride
Molecular Weight (g/mol) 222.69
Formula Weight 222.023
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.