missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenyl 2,3,4-Tri-O-acetyl-Beta-D-glucuronide Methyl Ester, TRC

Product Code. 30320051
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320051 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320051 Supplier Toronto Research Chemicals Supplier No. TRCA627200250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Aminophenyl 2,3,4-Tri-O-acetyl-beta-D-glucuronide Methyl Ester
CAS 25218-22-8
Molecular Formula C19 H23 N O10
Quantity 250 mg
Synonym 4-Aminophenyl 2,3,4-Tri-O-Acetyl-B-D-Glucuronide Methyl Ester
InChI Formula InChI=1S/C19H23NO10/c1-9(21)26-14-15(27-10(2)22)17(28-11(3)23)19(30-16(14)18(24)25-4)29-13-7-5-12(20)6-8-13/h5-8,14-17,19H,20H2,1-4H3/t14-,15-,16-,17+,19+/m0/s1
SMILES COC(=O)[C@H]1O[C@@H](Oc2ccc(N)cc2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-aminophenoxy)oxane-2-carboxylate
Molecular Weight (g/mol) 425.39
Formula Weight 425.1322
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.