missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalhydrazide, 98%

Product Code. 15436649
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436649 5 g 5g
15426649 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436649 Supplier Thermo Scientific Alfa Aesar Supplier No. J64921.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A chemiluminescent molecule | CAS: 3682-14-2 | C8H7N3O2 | 177.16 g/mol

A chemiluminescent molecule

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3682-14-2
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.16
MDL Number MFCD00010560
InChI Key HUDPLKWXRLNSPC-UHFFFAOYSA-N
Synonym 4-aminophthalhydrazide, isoluminol, 6-aminophthalazine-1,4-diol, chembl14882, 1,4-phthalazinedione, 6-amino-2,3-dihydro, 6-amino-2,3-dihydro-1,4-phthalazinedione, iso-luminol, 1enu, 4-aminophthaloylhydrazine, 4-amino-phthalsaeure-hydrazid
PubChem CID 95014
SMILES NC1=CC=C2C(=O)NNC(=O)C2=C1

Specifications

Melting Point 300°C
Color White to Yellow
Quantity 5 g
Beilstein 164804
Sensitivity Air Sensitive
Solubility Information Soluble in ammonium hydroxide,sodium carbonate or other base.
IUPAC Name 6-amino-2,3-dihydrophthalazine-1,4-dione
Formula Weight 177.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminophthalhydrazide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.