missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 4-Aminopyridine

Product Code. 18772494
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18772494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18772494 Supplier Tocris Bioscience™ Supplier No. 0940/100
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    129.15 € / 100mg
    Save 7.85 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Non-selective KV channel blocker

Non-selective voltage-dependent K+-channel blocker (IC50 values are 170 and 230 μM at KV1.1 and KV1.2 respectively). Caged 4-AP (Cat. No. 3557) is also available.
TRUSTED_SUSTAINABILITY

Specifications

CAS 504-24-5
Quantity 100 mg
Molecular Formula C5H6N2
Formula Weight 94.12
Purity >99%
Molecular Weight (g/mol) 94.117
Chemical Name or Material 4-Aminopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.