missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyrimidine, 98%

Product Code. 15403677
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403677 250 mg 250mg
11387139 1 g 1g
11397139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15403677 Supplier Thermo Scientific Alfa Aesar Supplier No. H27367.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 591-54-8 | C4H5N3 | 95.105 g/mol

4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminopyrimidine has been used in the preparation of 1:4-dihydro-4-imino-1-methylpyrimidine hydriodide.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-54-8
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD00006112
InChI Key OYRRZWATULMEPF-UHFFFAOYSA-N
Synonym 4-aminopyrimidine, 4-pyrimidinamine, 4-pyrimidineamine, pyrimidin-4-ylamine, pyrimidine, 4-amino, 6-aminopyrimidine, 4-amino pyrimidine, pyrimidine-4-ylamine, pyrimidin-4-yl-amine, 4-pyrimidinamine 9ci
PubChem CID 68958
ChEBI CHEBI:38616
IUPAC Name pyrimidin-4-amine
SMILES C1=CN=CN=C1N

Specifications

Melting Point 147°C to 152°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 95.11
Percent Purity 98%
Chemical Name or Material 4-Aminopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.