missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoquinoline, 95+%, Thermo Scientific™

Product Code. 15540654 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15540654 250 mg 250mg
15530654 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15540654 Supplier Thermo Scientific Acros Supplier No. 459762500
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 578-68-7
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.17
InChI Key FQYRLEXKXQRZDH-UHFFFAOYSA-N
Synonym 4-aminoquinoline, 4-quinolinamine, 4-aminoguinoline, quinolin-4-ylamine, 4-quinolylamine, unii-gte5p5l97n, ccris 1679, gte5p5l97n, 4-quinolinylamine, 4-amino quinoline
PubChem CID 68476
IUPAC Name quinolin-4-amine
SMILES C1=CC=C2C(=C1)C(=CC=N2)N

Specifications

Boiling Point 332.0°C to 337.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: slightly soluble.
Formula Weight 144.17
Percent Purity 95+%
Chemical Name or Material 4-Aminoquinoline
compliance-icons
Product Identifier
  • 4-Aminoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.