missing translation for 'onlineSavingsMsg'
Learn More

4-Anisonitrile, TRC

Product Code. 30348214
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348214 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348214 Supplier Toronto Research Chemicals Supplier No. TRCA67393510G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Anisonitrile
CAS 874-90-8
Molecular Formula C8H7NO
Quantity 10 g
Synonym 4-Methoxy-benzonitrile, 1-Cyano-4-methoxybenzene, 4-Cyanoanisole, Aubepinenitrile, p-Cyanoanisole, p-Cyanophenyl Methyl Ether, p-Methoxybenzonitrile, p-Methoxycyanobenzene, p-Methoxyphenyl Cyanide, NSC 3777, NSC 71539
InChI Formula InChI=1S/C8H7NO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,1H3
SMILES COc1ccc(cc1)C#N
IUPAC Name 4-methoxybenzonitrile
Molecular Weight (g/mol) 133.15
Formula Weight 133.0528
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.