missing translation for 'onlineSavingsMsg'
Learn More

4-Arsanilic Acid, TRC

Product Code. 30348675
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 30348675

Brand: Toronto Research Chemicals TRCA77411025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Arsanilic acid
CAS 98-50-0
Molecular Formula C6 H8 As N O3
Quantity 25 g
Synonym Arsanilic acid (8CI), Arsonic acid, (4-aminophenyl)- (9CI), As-(4-Aminophenyl)arsonic acid, (p-Aminophenyl)arsonic acid, 4-Aminobenzenearsonic acid, 4-Aminophenylarsonic acid, 4-Arsanilic acid, 4-Arsonoaniline, AS 101, Aminophenylarsine acid, Arsanilic Acid 100, Atoxylic acid, NSC 2023, Pro-gen, Pro-gen 227 Premix, Pro-gen 90, p-Aminobenzenearsonic acid, p-Anilinearsonic acid, p-Arsanilic acid, p-Arsonoaniline
InChI Formula InChI=1S/C6H8AsNO3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H,8H2,(H2,9,10,11)
SMILES Nc1ccc(cc1)[As](=O)(O)O
IUPAC Name (4-aminophenyl)arsonic acid
Molecular Weight (g/mol) 217.05
Formula Weight 216.972
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.