missing translation for 'onlineSavingsMsg'
Learn More

4-Azabenzimidazole, 98%

Product Code. 11426027
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426027 1 g 1g
11436027 5 g 5g
11446027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426027 Supplier Thermo Scientific Alfa Aesar Supplier No. B25051.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 273-21-2 | C6H5N3 | 119.13 g/mol

4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agent and strong acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273-21-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00005579,MFCD01646138
InChI Key GAMYYCRTACQSBR-UHFFFAOYSA-N
Synonym 3h-imidazo 4,5-b pyridine, 4-azabenzimidazole, 1h-imidazo 4,5-b pyridine, 1-deazapurine, 3,4-diazaindole, 7-azabenzimidazole, imidazopyridine, 4h-imidazo 4,5-b pyridine, pyrido 2,3-d imidazole, azabenzimidazole
PubChem CID 67504
ChEBI CHEBI:59952
IUPAC Name 1H-imidazo[4,5-b]pyridine
SMILES N1C=NC2=NC=CC=C12

Specifications

Melting Point 145°C to 150°C
Boiling Point 140°C to 150°C (0.1 mmHg)
Quantity 1 g
Beilstein 2600
Solubility Information Soluble in dimethylformamide.
Formula Weight 119.13
Percent Purity 99%
Chemical Name or Material 4-Azabenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.