missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-3-thiosemicarbazide, 98+%

Product Code. 11323076
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11323076

Brand: Thermo Scientific Alfa Aesar L10920.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13431-41-9 | C8H11N3S | 181.257 g/mol

4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13431-41-9
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.257
MDL Number MFCD00025145
InChI Key ZTRUHAVBRPABTK-UHFFFAOYSA-N
Synonym 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea
PubChem CID 737135
IUPAC Name 1-amino-3-benzylthiourea
SMILES C1=CC=C(C=C1)CNC(=S)NN

Specifications

Melting Point 128°C to 131°C
Quantity 5 g
UN Number UN2811
Beilstein 2096431
Solubility Information Insoluble in water.
Formula Weight 181.26
Percent Purity ≥98%
Chemical Name or Material 4-Benzyl-3-thiosemicarbazide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.