missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylmorpholine-2-carbonitrile, TRC

Product Code. 30342332
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30342332 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30342332 Supplier Toronto Research Chemicals Supplier No. TRCB53516050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Benzylmorpholine-2-carbonitrile
CAS 126645-52-1
Molecular Formula C12H14N2O
Quantity 50 mg
Synonym 4-(Phenylmethyl)-2-morpholinecarbonitrile, 2-Cyano-4-benzylmorpholine
InChI Formula InChI=1S/C12H14N2O/c13-8-12-10-14(6-7-15-12)9-11-4-2-1-3-5-11/h1-5,12H,6-7,9-10H2
SMILES N#CC1CN(Cc2ccccc2)CCO1
IUPAC Name 4-benzylmorpholine-2-carbonitrile
Molecular Weight (g/mol) 202.25
Formula Weight 202.1106
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.