missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxy-1-bromobenzene, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Benzyloxy-1-bromobenzene |
| CAS | 6793-92-6 |
| Molecular Formula | C13H11BrO |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C13H11BrO/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-9H,10H2 |
| SMILES | Brc1ccc(OCc2ccccc2)cc1 |
| IUPAC Name | 1-bromo-4-phenylmethoxybenzene |
| Molecular Weight (g/mol) | 263.13 |
| Formula Weight | 261.9993 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction