missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)-3-fluorophenylboronic acid, 97%, Varying anhydride amounts, Thermo Scientific™

Product Code. 10633413
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10633413 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10633413 Supplier Thermo Scientific Maybridge Supplier No. MO00842DA
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 133057-83-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H12BFO3
Linear Formula C6H5CH2OC6H3(F)B(OH)2
MDL Number MFCD00994627
Quantity 1 g
Synonym 4-benzyloxy-3-fluorophenylboronic acid, 3-fluoro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-fluorobenzeneboronic acid, 4-benzyloxy-3-fluorophenyl boronic acid, 4-benzyloxy-3-fluorophenyl boranediol, boronic acid, 3-fluoro-4-phenylmethoxy phenyl, pubchem5322
InChI Key DQPDLDQJMUGVGI-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)F)(O)O
IUPAC Name (3-fluoro-4-phenylmethoxyphenyl)boronic acid
Molecular Weight (g/mol) 246.044
PubChem CID 2737788
Formula Weight 246.04
Percent Purity 97%
Chemical Name or Material 4-(Benzyloxy)-3-fluorophenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.