missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)aniline Hydrochloride, TRC

Product Code. 30332094
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332094 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332094 Supplier Toronto Research Chemicals Supplier No. TRCB28549025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Benzyloxy)aniline Hydrochloride
CAS 51388-20-6
Molecular Formula C16 H20 F N3 O2 S
Quantity 25 g
Synonym 4-(Phenylmethoxy)benzeneamine Hydrochloride, 4-(Phenylmethoxy)aniline Hydrochloride, 4-Benzyloxyaniline Hydrochloric Acid Salt, 4-Benzyloxyphenylamine Hydrochloride, 4-[(Phenylmethyl)oxy]aniline Hydrochloride, p-(Benzyloxy)aniline Hydrochloride
InChI Formula InChI=1S/C16H20FN3O2S/c1-10(2)14-11(3)15(12-6-8-13(17)9-7-12)19-16(18-14)20(4)23(5,21)22/h6-10H,1-5H3
SMILES CC(C)c1nc(nc(c1C)c2ccc(F)cc2)N(C)S(=O)(=O)C
IUPAC Name N-[4-(4-fluorophenyl)-5-methyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
Molecular Weight (g/mol) 337.412
Formula Weight 337.126
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.