missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)phenyl isothiocyanate 97%, Thermo Scientific™

Product Code. 10458114
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10458114 5 g 5g
10109662 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10458114 Supplier Thermo Scientific Maybridge Supplier No. SB01753DE
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 139768-71-1
Molecular Formula C14H11NOS
Molecular Weight (g/mol) 241.308
InChI Key OQXRBXAFSXVCCO-UHFFFAOYSA-N
Synonym 4-benzyloxyphenyl isothiocyanate, 4-benzyloxyphenylisothiocyanate, 1-benzyloxy-4-isothiocyanatobenzene, 4-phenylmethoxy benzenisothiocyanate, acmc-20amoj, 4-benzyloxyisothiocyanate, 4-phenylmethoxyphenylisothiocyanate, 4-benzyloxy phenyl isothiocyanate, 4-phenylmethoxy phenyl isothiocyanate, 1-isothiocyanato-4-phenylmethoxy-benzene
PubChem CID 2735503
IUPAC Name 1-isothiocyanato-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)N=C=S

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 241.31
Percent Purity 97%
Chemical Name or Material 4-(Benzyloxy)phenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.