missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyaniline hydrochloride, 98%

Product Code. 10744091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10744091 100 g Plastic bottle 100g
10734091 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10744091 Supplier Thermo Scientific Acros Supplier No. 152291000
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 51388-20-6 | C13H13NO·HCl | 235.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51388-20-6
Molecular Formula C13H13NO·HCl
Molecular Weight (g/mol) 235.72
MDL Number MFCD00012995
InChI Key KQBDLOVXZHOAJI-UHFFFAOYSA-N
Synonym 4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343
PubChem CID 2723831
IUPAC Name 4-phenylmethoxyaniline;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 228°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5CH2OC6H4NH2·HCl
Quantity 100 g
Beilstein 13, 439
Solubility Information Solubility in water: slightly soluble
Formula Weight 235.72
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxyaniline hydrochloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Benzyloxyaniline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.