missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyaniline hydrochloride, 98%

Product Code. 11492878
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492878 25 g 25g
11402888 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492878 Supplier Thermo Scientific Alfa Aesar Supplier No. A16856.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51388-20-6 | C13H14ClNO | 235.711 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51388-20-6
Molecular Formula C13H14ClNO
Molecular Weight (g/mol) 235.711
MDL Number MFCD00012995
InChI Key KQBDLOVXZHOAJI-UHFFFAOYSA-N
Synonym 4-benzyloxyaniline hydrochloride, 4-benzyloxy aniline hydrochloride, 4-benzyloxyaniline hcl, benzenamine, 4-phenylmethoxy-, hydrochloride, 4-phenylmethoxyaniline hydrochloride, aniline, p-benzyloxy-hydrochloride, 4-benzyloxy phenyl amine hydrochloride, 4-phenylmethoxy phenylamine, chloride, acmc-20amop, pubchem21343
PubChem CID 2723831
IUPAC Name 4-phenylmethoxyaniline;hydrochloride
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)N.Cl

Specifications

Melting Point ∼225°C (decomposition)
Quantity 25 g
Beilstein 3633307
Formula Weight 235.72
Percent Purity 98%
Chemical Name or Material 4-Benzyloxyaniline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.