missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxyphenyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11362407
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362407 5 g 5g
11352407 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362407 Supplier Thermo Scientific Alfa Aesar Supplier No. L10038.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Hydrolyzes in water.

Notes
Air and moisture sensitive. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139768-71-1
Molecular Formula C14H11NOS
Molecular Weight (g/mol) 241.308
MDL Number MFCD00041095
InChI Key OQXRBXAFSXVCCO-UHFFFAOYSA-N
Synonym 4-benzyloxyphenyl isothiocyanate, 4-benzyloxyphenylisothiocyanate, 1-benzyloxy-4-isothiocyanatobenzene, 4-phenylmethoxy benzenisothiocyanate, acmc-20amoj, 4-benzyloxyisothiocyanate, 4-phenylmethoxyphenylisothiocyanate, 4-benzyloxy phenyl isothiocyanate, 4-phenylmethoxy phenyl isothiocyanate, 1-isothiocyanato-4-phenylmethoxy-benzene
PubChem CID 2735503
IUPAC Name 1-isothiocyanato-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)N=C=S

Specifications

Melting Point 62°C to 65°C
Quantity 5 g
UN Number UN2923
Beilstein 3298091
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 241.31
Percent Purity 98%
Chemical Name or Material 4-Benzyloxyphenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.