missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylboronic acid, 97%

Product Code. 10058139 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10058139 5 g Glass bottle 5g
10637210 1 g Glass bottle 1g
10068139 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10058139 Supplier Thermo Scientific Acros Supplier No. 394420050
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5122-94-1 | C12H11BO2 | 198.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 5122-94-1
Molecular Formula C12H11BO2
Molecular Weight (g/mol) 198.03
MDL Number MFCD00093311
InChI Key XPEIJWZLPWNNOK-UHFFFAOYSA-N
Synonym 4-biphenylboronic acid, 1,1'-biphenyl-4-ylboronic acid, biphenyl-4-boronic acid, 4-biphenyl boronic acid, biphenylboronic acid, 4-phenylphenyl boranediol, 4-phenylphenyl boronic acid, biphenyl-4-ylboronic acid, 1,1'-biphenyl-4-yl-boronic acid, 4-phenylbenzeneboronic acid
PubChem CID 151253
IUPAC Name (4-phenylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C1=CC=CC=C1

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point 241.0°C to 245.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5C6H4B(OH)2
Quantity 5 g
Formula Weight 198.03
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Biphenylboronic acid
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 4-Biphenylboronic acid
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.