missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylcarbonyl chloride, 98%

Product Code. 10154090 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10154090 10 g 10g
10409460 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10154090 Supplier Thermo Scientific Acros Supplier No. 171250100
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 14002-51-8 | C13H9ClO | 216.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14002-51-8
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000692
InChI Key JPVUWCPKMYXOKW-UHFFFAOYSA-N
Synonym 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride
PubChem CID 84151
IUPAC Name 4-phenylbenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 110°C to 114°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula C6H5C6H4COCl
Quantity 10 g
Beilstein 09, I, 280
Fieser 04,376; 07,282
Solubility Information Solubility in water: reacts. Other solubilities: soluble in chloroform,dmf
Formula Weight 216.67
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Biphenylcarbonyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Biphenylcarbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.