missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%

Product Code. 15441528
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441528 5 g 5g
15431528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441528 Supplier Thermo Scientific Alfa Aesar Supplier No. H55311.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153747-97-8 | C14H20BrN3O2 | 342.237 g/mol

4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153747-97-8
Molecular Formula C14H20BrN3O2
Molecular Weight (g/mol) 342.237
MDL Number MFCD07369772
InChI Key DSLVSFMWCDGZIL-UHFFFAOYSA-N
Synonym tert-butyl 4-5-bromopyridin-2-yl piperazine-1-carboxylate, 2-4-boc-piperazino-5-bromopyridine, 4-boc-1-5-bromo-2-pyridyl piperazine, 5-bromo-2-4-n-boc piperazin-1-yl pyridine, 4-5-bromo-pyridin-2-yl-piperazine-1-carboxylic acid tert-butyl ester, 5-bromo-2-4-tert-butoxycarbonylpiperazin-1-yl pyridine, 4-5-bromopyridin-2-yl piperazine-1-carboxylic acid tert-butyl ester, 1-piperazinecarboxylic acid, 4-5-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ah70, 2-n-boc-piperazin-1-yl-5-bromopyridine
PubChem CID 11244775
IUPAC Name tert-butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)Br

Specifications

Melting Point 83°C to 87°C
Color White to Yellow
Quantity 5 g
UN Number UN2811
Solubility Information Soluble in water.
Formula Weight 342.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Boc-1-(5-bromo-2-pyridyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.