missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(6-methyl-2-pyridyl)piperazine, 97%, Thermo Scientific™

Product Code. 15452208
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
15452208 1 g 1g
15462208 5 g 5g
15472208 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 15452208

Brand: Thermo Scientific Alfa Aesar H55841.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127188-33-4
Molecular Formula C15H23N3O2
Molecular Weight (g/mol) 277.368
MDL Number MFCD07369771
InChI Key QEDVZUFNZJJSJL-UHFFFAOYSA-N
Synonym 4-boc-1-6-methyl-2-pyridyl piperazine, tert-butyl 4-6-methylpyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1c1la, 4-boc-1-6-methylpyridin-2-yl piperazine, tert-butyl-4-6-methylpyridin-2-yl piperazine-1-carboxylate
PubChem CID 16217938
IUPAC Name tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
SMILES CC1=NC(=CC=C1)N2CCN(CC2)C(=O)OC(C)(C)C

Specifications

Melting Point 80°C to 84°C
Color White
Quantity 1 g
UN Number UN2811
Formula Weight 277.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Boc-1-(6-methyl-2-pyridyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.