missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)benzeneboronic acid, 95%

Product Code. 15499977
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499977 250 mg 250mg
15489977 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499977 Supplier Thermo Scientific Alfa Aesar Supplier No. H53225.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 489446-42-6 | C12H18BNO4 | 251.09 g/mol

4-(Boc-aminomethyl)benzeneboronic acid is a boc protected phenyl boronic acid used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Boc-aminomethyl)benzeneboronic acid is a boc protected phenyl boronic acid used for proteomics research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 489446-42-6
Molecular Formula C12H18BNO4
Molecular Weight (g/mol) 251.09
MDL Number MFCD04115637
InChI Key MUBGEKQUCSEECZ-UHFFFAOYSA-N
Synonym 4-n-boc-aminomethyl phenylboronic acid, 4-n-boc-amino methyl phenylboronic acid, 4-tert-butoxycarbonyl amino methyl phenyl boronic acid, 4-n-boc aminomethylphenylboronic acid, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, 4-2-methylpropan-2-yl oxycarbonylamino methyl phenyl boronic acid, 4-boc-aminomethyl benzeneboronic acid, 4-aminomethyl benzeneboronic acid, n-boc protected, 4-tert-butoxycarbonylamino methyl phenylboronic acid
PubChem CID 3863163
IUPAC Name [4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]boronic acid
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 204°C to 206°C
Quantity 250 mg
Formula Weight 251.09
Percent Purity 97%
Chemical Name or Material 4-(Boc-aminomethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.