missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-(methylenedioxy)benzene, 97%

Product Code. 10115660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10115660 25 g Glass bottle 25g
10117713 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10115660 Supplier Thermo Scientific Acros Supplier No. 307680250
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2635-13-4 | C7H5BrO2 | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2635-13-4
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00005821
InChI Key FBOYMIDCHINJKC-UHFFFAOYSA-N
Synonym 4-bromo-1,2-methylenedioxy benzene, 5-bromobenzo d 1,3 dioxole, 4-bromo-1,2-methylenedioxybenzene, 1-bromo-3,4-methylenedioxy benzene, 3,4-methylenedioxybromobenzene, 5-bromo-2h-1,3-benzodioxole, 5-bromobenzo-1,3-dioxole, 5-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo, 3,4-methylenedioxy bromobenzene
PubChem CID 75831
SMILES BrC1=CC=C2OCOC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Orange-Yellow
Density 1.6600g/mL
Boiling Point 85°C to 86°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5820 to 1.584
Quantity 25 g
Specific Gravity 1.66
IUPAC Name 5-bromo-2H-1,3-benzodioxole
Formula Weight 201.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1, 2-(methylenedioxy)benzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.