missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butanol, tech., cont. varying amounts of THF

Product Code. 11397218
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397218 1 g 1g
11387218 5 g 5g
11377218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11397218 Supplier Thermo Scientific Alfa Aesar Supplier No. 016805.03
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33036-62-3 | C4H9BrO | 153.019 g/mol

4-Bromo-1-butanol is used to prepare 2-(4-bromo-butoxy)-tetrahydro-pyran by reacting with 3,4-dihydro-2H-pyran in the presence of amberlyst 15 as a catalyst. It reacts with dihydropyran to prepare tetrahydropyranyl ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33036-62-3
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.019
MDL Number MFCD00015387
InChI Key SIJLYRDVTMMSIP-UHFFFAOYSA-N
Synonym 4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv
PubChem CID 118388
IUPAC Name 4-bromobutan-1-ol
SMILES C(CCBr)CO

Specifications

Color Yellow
Boiling Point 63°C to 65°C (5 mmHg)
Flash Point 32°C (91°F)
Linear Formula Br(CH2)4OH
Refractive Index 1.5035
Quantity 1 g
UN Number UN1987
Sensitivity Light and moisture sensitive
Solubility Information Immiscible with water.
Formula Weight 153.02
Grade Technical
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butanol, cont. varying amounts of THF
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.