missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazol-3-amine, Tech., Thermo Scientific™

Product Code. 10357064
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10357064 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10357064 Supplier Thermo Scientific Maybridge Supplier No. GK00323DA
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 146941-72-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C4H6BrN3
MDL Number MFCD00663065
Quantity 1 g
Synonym 4-bromo-1-methyl-1h-pyrazol-3-amine, 3-amino-4-bromo-1-methyl-1h-pyrazole, 3-amino-4-bromo-1-methylpyrazole, 1h-pyrazol-3-amine, 4-bromo-1-methyl, 4-bromo-1-methylpyrazole-3-ylamine, acmc-1bzbh, 1h-pyrazol-3-amine,4-bromo-1-methyl
InChI Key WYPUMGAZSVZUGJ-UHFFFAOYSA-N
SMILES CN1C=C(Br)C(N)=N1
IUPAC Name 4-bromo-1-methylpyrazol-3-amine
Molecular Weight (g/mol) 176.02
PubChem CID 2735283
Formula Weight 176.02
Percent Purity 97%
Grade Technical
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazol-3-amine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.