missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific Chemicals

Product Code. 15485927
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485927 1 g 1g
15428335 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485927 Supplier Thermo Scientific Alfa Aesar Supplier No. H33131.03
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 287922-71-8 | Molecular Formula: C5H4BrN3 | Molecular Weight: 186.01 g/mol

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287922-71-8
Molecular Formula C5H4BrN3
Molecular Weight (g/mol) 186.01
MDL Number MFCD00103545
InChI Key NTTLCOOFUGHMJS-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carbonitrile, 1h-pyrazole-3-carbonitrile, 4-bromo-1-methyl, 4-bromo-1-methyl-pyrazole-3-carbonitrile
PubChem CID 45594322
IUPAC Name 4-bromo-1-methylpyrazole-3-carbonitrile
SMILES CN1C=C(Br)C(=N1)C#N

Specifications

Melting Point 103°C to 105°C
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 186.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.