missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole, 98+%

Product Code. 15442109
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15442109 100 g 100g
15432109 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442109 Supplier Thermo Scientific Alfa Aesar Supplier No. H64768.22
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15803-02-8 | C4H5BrN2 | 161.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15803-02-8
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.00
MDL Number MFCD02179565
InChI Key IXJSDKIJPVSPKF-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole, 1-methyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-1-methyl, 1-mehtyl-4-bromo-1h-pyrazole, 1-methyl-4-bromopyrazole, 4-bromo-1-methyl-pyrazole, 4-bromo-2-methyl-1h-pyrazole, zlchem 151, pubchem16867, 4bmp
PubChem CID 167433
IUPAC Name 4-bromo-1-methylpyrazole
SMILES CN1C=C(Br)C=N1

Specifications

Color Colorless to Yellow
Density 1.558 g/mL
Boiling Point 185°C to 188°C
Flash Point 94°C (201°F)
Refractive Index 1.531
Quantity 100 g
Beilstein 108120
Formula Weight 161
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.