missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-naphthoic acid, TRC

Product Code. 30345944
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30345944 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30345944

Brand: Toronto Research Chemicals TRCB697935500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromo-1-naphthoic acid
CAS 16650-55-8
Molecular Formula C11H7BrO2
Quantity 500 mg
Synonym 4-Bromo-1-Naphthalenecarboxylic Acid
InChI Formula InChI=1S/C11H7BrO2/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H,(H,13,14)
SMILES OC(=O)c1ccc(Br)c2ccccc12
IUPAC Name 4-bromonaphthalene-1-carboxylic acid
Molecular Weight (g/mol) 251.08
Formula Weight 249.9629
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.