missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-(triisopropylsilyl)-1H-indole, ≥97%, Thermo Scientific™

Product Code. 10699554
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10699554 250 mg Amber Glass Bottle 250mg
10577863 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10699554 Supplier Thermo Scientific Maybridge Supplier No. CC39510CB
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 412048-44-3
Molecular Formula C17H26BrNSi
Molecular Weight (g/mol) 352.391
MDL Number MFCD05664418
InChI Key ZXHMUQVMGJYGAV-UHFFFAOYSA-N
Synonym 4-bromo-1-triisopropylsilyl-1h-indole, 4-bromo-1-triisopropylsilyl indole, 4-bromo-triisopropylsilanyl-1h-indole, 4-bromo-1-triisopropylsilanyl-1h-indole, 4-bromo-1-tri-isopropylsilyl-1h-indole, 4-bromoindol-1-yl-tri propan-2-yl silane
PubChem CID 2795573
IUPAC Name (4-bromoindol-1-yl)-tri(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=C1C=CC=C2Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 352.39
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-(triisopropylsilyl)-1H-indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.