missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific™

Product Code. 10663223
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10663223 1 g Amber Glass Bottle 1g
10273612 250 mg Amber Glass Bottle 250mg
10317354 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10663223 Supplier Thermo Scientific Maybridge Supplier No. RF05036DA
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 288246-16-2
Molecular Formula C4H2BrN3
Molecular Weight (g/mol) 171.985
MDL Number MFCD00105683
InChI Key ZXVFKQRZKKGVNJ-UHFFFAOYSA-N
Synonym 4-bromo-1h-pyrazole-3-carbonitrile, 4-bromo-2h-pyrazole-3-carbonitrile, 4-bromopyrazole-3-carbonitrile, 4-bromo-3-cyano-1h-pyrazole, 1h-pyrazole-3-carbonitrile, 4-bromo, pubchem10182, acmc-20aa8h, maybridge1_006470, ksc497k2j, buttpark 181\04-37
PubChem CID 2735623
IUPAC Name 4-bromo-1H-pyrazole-5-carbonitrile
SMILES C1=NNC(=C1Br)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 171.98
Percent Purity 97%
Chemical Name or Material 4-Bromo-1H-pyrazole-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.