missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,2-difluoro-1,3-benzodioxole, 97%

Product Code. 11475827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475827 5 g 5g
11465827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475827 Supplier Thermo Scientific Alfa Aesar Supplier No. B24389.06
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 144584-66-7 | C7H3BrF2O2 | 237 g/mol

4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
It is unstable under normal temperature. storage temperature is 2 - 4°C. Incompatible material is oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144584-66-7
Molecular Formula C7H3BrF2O2
Molecular Weight (g/mol) 237
MDL Number MFCD01631385
InChI Key LSZYHXNOLVSZHH-UHFFFAOYSA-N
Synonym 4-bromo-2,2-difluorobenzo d 1,3 dioxole, 4-bromo-2,2-difluoro-2h-1,3-benzodioxole, 2,2-difluoro-4-bromo-1,3-benzodioxole, 1,3-benzodioxol-4-bromo, 2,2-difluoro, 4-bromo-2,2-difluoro-benzo 1,3 dioxole, 4-bromo-2,2-difluorobenzo d 1,3-dioxolene, acmc-20a3ue, 4-bromo-2,2-difluoro-1,3-benzodioxol, 1,3-benzodioxole,4-bromo-2,2-difluoro, 4-bromo-2,2-difluoro-1,3-benzodioxolane
PubChem CID 2773297
IUPAC Name 4-bromo-2,2-difluoro-1,3-benzodioxole
SMILES C1=CC2=C(C(=C1)Br)OC(O2)(F)F

Specifications

Refractive Index 1.4968
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 237
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,2-difluoro-1,3-benzodioxole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.