missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2,5-dichlorothiophene-3-sulfonyl chloride, 97%

Product Code. 11434204
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434204 1 g 1g
11454204 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434204 Supplier Alfa Aesar™ Supplier No. A12533.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 166964-36-9
Molecular Formula C4BrCl3O2S2
Molecular Weight (g/mol) 330.416
MDL Number MFCD00051664
InChI Key NAHZODWKWDZKSZ-UHFFFAOYSA-N
Synonym 4-bromo-2,5-dichlorothiophene-3-sulphonyl chloride, acmc-20amnx, 4-bromo-2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride,4-bromo-2,5-dichloro, 4-bromo-2,5-dichloro-thiophene-3-sulfonylchloride, 3-thiophenesulfonylchloride, 4-bromo-2,5-dichloro, 4-bromo-2,5-dichloro-3-thiophenesulfonyl chloride #
PubChem CID 519295
IUPAC Name 4-bromo-2,5-dichlorothiophene-3-sulfonyl chloride
SMILES C1(=C(SC(=C1Br)Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 67°C to 71°C
Boiling Point 119°C to 121°C (1 mmHg)
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 330.44
Percent Purity 97%
Chemical Name or Material 4-Bromo-2,5-dichlorothiophene-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.