missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-dichlorothiophene-3-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10480554
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10480554 25 g 25g
10405892 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10480554 Supplier Thermo Scientific Maybridge Supplier No. CD09555EB
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 166964-36-9
Molecular Formula C4BrCl3O2S2
Molecular Weight (g/mol) 330.416
MDL Number MFCD00051664
InChI Key NAHZODWKWDZKSZ-UHFFFAOYSA-N
Synonym 4-bromo-2,5-dichlorothiophene-3-sulphonyl chloride, acmc-20amnx, 4-bromo-2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonylchloride,4-bromo-2,5-dichloro, 4-bromo-2,5-dichloro-thiophene-3-sulfonylchloride, 3-thiophenesulfonylchloride, 4-bromo-2,5-dichloro, 4-bromo-2,5-dichloro-3-thiophenesulfonyl chloride #
PubChem CID 519295
IUPAC Name 4-bromo-2,5-dichlorothiophene-3-sulfonyl chloride
SMILES C1(=C(SC(=C1Br)Cl)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 330.44
Percent Purity 97%
Chemical Name or Material 4-Bromo-2, 5-dichlorothiophene-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.