missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzenesulfonyl chloride, 96%

Product Code. 11441397
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441397 5 g 5g
11451397 25 g 25g
11461397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441397 Supplier Thermo Scientific Alfa Aesar Supplier No. B24913.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 351003-52-6 | C6H3BrCl2O2S | 289.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-52-6
Molecular Formula C6H3BrCl2O2S
Molecular Weight (g/mol) 289.95
MDL Number MFCD03094630
InChI Key OECYEHWJPRPCOH-UHFFFAOYSA-N
Synonym 4-bromo-2-chlorobenzene-1-sulfonyl chloride, 2-chloro-4-bromo-benzenesulfonyl chloride, 4-bromo-2-chlorobenzenesulphonyl chloride, 4-bromo-2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 4-bromo-2-chloro, 4-bromo-2-chlorophenylsulfonyl chloride, pubchem11732, 4-bromo-2-chloro-benzenesulfonyl chloride, acmc-1ag54, ksc221s9p
PubChem CID 2734766
IUPAC Name 4-bromo-2-chlorobenzenesulfonyl chloride
SMILES ClC1=CC(Br)=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 76°C to 81°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 289.96
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.