missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzenesulfonyl chloride, 97%, ACROS Organics™

Product Code. 10734183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10734183 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10734183 Supplier Thermo Scientific Acros Supplier No. 371430010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 351003-52-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 75°C to 80°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H3BrCl2O2S
Linear Formula BrC6H3ClSO2Cl
MDL Number MFCD03094630
Quantity 1 g
Synonym 4-bromo-2-chlorobenzene-1-sulfonyl chloride, 2-chloro-4-bromo-benzenesulfonyl chloride, 4-bromo-2-chlorobenzenesulphonyl chloride, 4-bromo-2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 4-bromo-2-chloro, 4-bromo-2-chlorophenylsulfonyl chloride, pubchem11732, 4-bromo-2-chloro-benzenesulfonyl chloride, acmc-1ag54, ksc221s9p
InChI Key OECYEHWJPRPCOH-UHFFFAOYSA-N
SMILES ClC1=CC(Br)=CC=C1S(Cl)(=O)=O
IUPAC Name 4-bromo-2-chlorobenzene-1-sulfonyl chloride
Molecular Weight (g/mol) 289.95
PubChem CID 2734766
Formula Weight 289.96
Percent Purity ≥96.0%
Physical Form Crystals and/or Chunks
Chemical Name or Material 4-Bromo-2-chlorobenzenesulfonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-chlorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.