missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-chlorophenol, TRC
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-chlorophenol |
| CAS | 3964-56-5 |
| Molecular Formula | C6 H4 Br Cl O |
| Quantity | 1 g |
| Synonym | 4-Bromo-2-chlorophenol, 2-Chloro-4-bromophenol |
| InChI Formula | InChI=1S/C6H4BrClO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H |
| SMILES | Oc1ccc(Br)cc1Cl |
| IUPAC Name | 4-bromo-2-chlorophenol |
| Molecular Weight (g/mol) | 207.45 |
| Formula Weight | 205.9134 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction