missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorotoluene, 98%, Thermo Scientific™

Product Code. 10759585 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10759585 100 g Glass bottle 100g
10400934 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10759585 Supplier Thermo Scientific Acros Supplier No. 398281000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 89794-02-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00060651
InChI Key LIFMTDJMLRECMX-UHFFFAOYSA-N
Synonym 4-bromo-2-chlorotoluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, benzene, 4-bromo-2-chloro-1-methyl, 4-bromo-chlorotoluene, pubchem3603, 4-bromo-2chlorotoluene, 2-chloro 4-bromo toluene, acmc-209r2x
PubChem CID 2735554
IUPAC Name 4-bromo-2-chloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Cl

Specifications

CAS Min % 97.5
Color Yellow
CAS Max % 100.0
Density 1.5380g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5730 to 1.5750 (20°C, 589nm)
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.538
Linear Formula BrC6H3(Cl)CH3
Boiling Point 212°C
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chlorotoluene
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-chlorotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.