missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzenesulfonyl chloride, 98%

Product Code. 11425787
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425787 5 g 5g
11415787 1 g 1g
11435787 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425787 Supplier Thermo Scientific Alfa Aesar Supplier No. B24352.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 216159-03-4 | C6H3BrClFO2S | 273.5 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216159-03-4
Molecular Formula C6H3BrClFO2S
Molecular Weight (g/mol) 273.5
MDL Number MFCD01090993
InChI Key XNYBZLRIUHNRQY-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzene-1-sulfonyl chloride, 2-fluoro-4-bromobenzenesulfonyl chloride, 4-bromo-2-fluorobenzenesulphonyl chloride, 4-bromo-2-fluoro-benzenesulfonyl chloride, buttpark 147\11-41, 4-bromo-2-fluorophenyl chlorosulfone, benzenesulfonyl chloride, 4-bromo-2-fluoro, pubchem5065, acmc-209via, ksc201o8l
PubChem CID 2736310
IUPAC Name 4-bromo-2-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Br)F)S(=O)(=O)Cl

Specifications

Melting Point 63°C to 66°C
Boiling Point 84°C (0.5 mmHg)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 273.51
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.