missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(methoxymethyl)pyridine, TRC

Product Code. 30422575
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422575 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422575 Supplier Toronto Research Chemicals Supplier No. TRCP994203100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromo-2-(methoxymethyl)pyridine
CAS 864412-04-4
Molecular Formula C7H8NOBr
Quantity 100 mg
InChI Formula InChI=1S/C7H8BrNO/c1-10-5-7-4-6(8)2-3-9-7/h2-4H,5H2,1H3
SMILES COCc1cc(Br)ccn1
IUPAC Name 4-bromo-2-(methoxymethyl)pyridine
Molecular Weight (g/mol) 202.05
Formula Weight 200.9789
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.