missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 15204904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15204904 10 g 10g
15214904 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15204904 Supplier Thermo Scientific Acros Supplier No. 451780100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 139937-37-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.55
InChI Key KNBSFUSBZNMAKU-UHFFFAOYSA-N
Synonym 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072
PubChem CID 2781338
IUPAC Name 4-bromo-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 62.0°C to 67.0°C
Boiling Point 164.0°C (15.0 mmHg)
Assay Percent Range 96% min. (GC)
Linear Formula Br(CH3)C6H3SO2Cl
Quantity 10 g
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzene-1-sulfonyl chloride
compliance-icons
Product Identifier
  • 4-Bromo-2-methylbenzene-1-sulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.