missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzenesulfonyl chloride, 97%

Product Code. 15494477
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494477 10 g 10g
15484477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494477 Supplier Thermo Scientific Alfa Aesar Supplier No. H31835.09
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 139937-37-4 | C7H6BrClO2S | 269.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139937-37-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD01365820
InChI Key KNBSFUSBZNMAKU-UHFFFAOYSA-N
Synonym 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072
PubChem CID 2781338
IUPAC Name 4-bromo-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 63°C to 65°C
Boiling Point 164°C (15 mmHg)
Quantity 10 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.