missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitroaniline, 97%, Thermo Scientific™

Product Code. 10406282
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10406282 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10406282 Supplier Thermo Scientific Maybridge Supplier No. KM06414DA
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 875-51-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H5BrN2O2
MDL Number MFCD00041312
Quantity 1 g
Synonym benzenamine, 4-bromo-2-nitro, 2-nitro-4-bromoaniline, 4-bromo-o-nitroaniline, p-bromo-o-nitroaniline, 4-bromo-2-nitro-phenylamine, 4-bromo-2-nitrobenzeneamine, aniline, 4-bromo-2-nitro, 4-bromo-2-nitrobenzenamine, 1-amino-4-bromo-2-nitrobenzene, 4-bromo-2-nitro-aniline
InChI Key ZCWBZRBJSPWUPG-UHFFFAOYSA-N
SMILES NC1=CC=C(Br)C=C1[N+]([O-])=O
IUPAC Name 4-bromo-2-nitroaniline
Molecular Weight (g/mol) 217.02
PubChem CID 70132
Formula Weight 217.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-nitroaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.