missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10661693
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10661693 10 g Amber Glass Bottle 10g
10140752 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10661693 Supplier Thermo Scientific Maybridge Supplier No. TL00664EA
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-14-5
Molecular Formula C7H3BrClF3O3S
Molecular Weight (g/mol) 339.51
MDL Number MFCD00179336
InChI Key ZZCIRIWJHAZTIW-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethoxy benzene-1-sulfonyl chloride, 4-bromo-2-trifluoromethoxy benzenesulfonyl chloride, 4-bromo-2-trifluoromethoxy benzene sulphonyl chloride, 4-bromo-2-trifluoromethoxy benzenesulfonylchloride, 4-bromo-2-trifluoromethoxy benzene, 4-bromo-2-trifluoromethoxy phenyl chlorosulfone, 4-bromo-2-trifluoromethoxy-benzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-trifluoromethoxy, benzenesulfonyl chloride, 4-bromo-2-trifluoromethoxy, 4-bromo-2-trifluoromethoxy benzenesulphonyl chloride
PubChem CID 2736421
IUPAC Name 4-bromo-2-(trifluoromethoxy)benzenesulfonyl chloride
SMILES FC(F)(F)OC1=C(C=CC(Br)=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Density 1.85g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 339.51
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(trifluoromethoxy)benzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.